Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United Kingdom
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
6-(Allyloxy)-2-chloro-3-fluorobenzyl Alcohol
6-(Allyloxy)-2-chloro-3-fluorobenzyl Alcohol
6-(Allyloxy)-2-chloro-3-fluorobenzyl Alcohol
APOSPC510152-1G
:
Apollo Scientific
:
6-(Allyloxy)-2-chloro-3-fluorobenzyl Alcohol
APOSPC510152-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
216.637 g/mol
:
Ambient
:
1823365-87-2
:
C₁₀H₁₀ClFO₂
:
MFCD26937567