


2-bromo-3-fluorophenylthiocyanate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:2149597-43-1
:
Formula:C₇H₃BrFNS
MDL Number:MFCD30723421
:232.073 g/mol
Storage Temperature:Ambient
UN:1759
ADR:8,III
:GHS05
:GHS07
:GHS08
Frequently Bought Together