Order Entry
United Kingdom
ContactUsLinkComponent
2,6-Difluoro-4-(trifluoromethyl)phenyl 5-fluoro-2-iodophenyl ether 95%
2,6-Difluoro-4-(trifluoromethyl)phenyl 5-fluoro-2-iodophenyl ether 95%
Catalog # APOSPC502816-1G
Supplier:  Apollo Scientific
CAS Number:  
2,6-Difluoro-4-(trifluoromethyl)phenyl 5-fluoro-2-iodophenyl ether 95%
Catalog # APOSPC502816-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :418.073 g/mol
Storage Temperature:Ambient
CAS Number:2244087-01-0
Formula:C₁₃H₅F₆IO
MDL Number:MFCD31630072