2,6-Difluoro-4-(trifluoromethyl)phenyl 4-fluoro-3-methylphenyl ether 95%
Catalog # APOSPC502808-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:306.203 g/mol
Storage Temperature:Ambient
CAS Number:2244086-25-5
Formula:C₁₄H₈F₆O
MDL Number:MFCD31630048