2,6-Difluoro-4-(trifluoromethyl)phenyl 5-iodo-2-nitrophenyl ether 95%
Catalog # APOSPC502763-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:445.08 g/mol
Storage Temperature:Ambient
CAS Number:2244088-49-9
Formula:C₁₃H₅F₅INO₃
MDL Number:MFCD31630062