Order Entry
United Kingdom
ContactUsLinkComponent
2-Bromo-5-fluoro-4-nitrophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
2-Bromo-5-fluoro-4-nitrophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
Catalog # APOSPC502758-1G
Supplier:  Apollo Scientific
CAS Number:  
2-Bromo-5-fluoro-4-nitrophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
Catalog # APOSPC502758-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :416.07 g/mol
Storage Temperature:Ambient
CAS Number:2244087-50-9
Formula:C₁₃H₄BrF₆NO₃
MDL Number:MFCD31630047