4-Bromo-3-fluoro-2-hydroxybenzaldehyde
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: 4-Bromo-3-fluorosalicylaldehyde, 3-Bromo-2-fluoro-6-formylphenol
CAS Number:1427373-29-2
:
Formula:C₇H₄BrFO₂
MDL Number:MFCD23705524
Molecular Weight:219.008 g/mol
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together