Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United Kingdom
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Chloro-6-(trifluoromethyl)phenol
2-Chloro-6-(trifluoromethyl)phenol
2-Chloro-6-(trifluoromethyl)phenol
2-Chloro-6-(trifluoromethyl)phenol
APOSPC500170-1G
:
Apollo Scientific
:
2-Chloro-6-(trifluoromethyl)phenol
APOSPC500170-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
Environmentally Preferable:
:
2-Trifluoromethyl-6-chlorophenol
:
:
196.554 g/mol
:
Ambient
:
106877-36-5
:
C₇H₄ClF₃O
:
MFCD00673986