2-{2-[3-Chloro-5-(trifluoromethyl)pyridin-2-ylamino]ethyl}amino-5-(trifluoromethyl)aniline hydrochloride
Catalog # APOSPC4915-500MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:500 mg
- Seq:2725
Specifications
About this item
:
:435.195 g/mol
Storage Temperature:Ambient
CAS Number:317822-50-7
Formula:C₁₅H₁₃ClF₆N₄·ClH
MDL Number:MFCD00793061