
2-Bromo-3-fluoro-6-nitrophenol 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:103979-08-4
:
Formula:C₆H₃BrFNO₃
MDL Number:MFCD18071060
:235.995 g/mol
Melting Pt:65…66 °C
Storage Temperature:Ambient
UN:2811
ADR:6.1,III
:GHS06
Frequently Bought Together