4,4,5,5-Tetramethyl-2-[2,3,5,6-tetrafluoro-4-(trifluoromethyl)phenyl]-1,3,2-dioxaborolane
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1111096-06-0
:
Formula:C₁₃H₁₂BF₇O₂
MDL Number:MFCD12405355
:344.033 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together