
1-Iodo-4-[1,1,2-trifluoro-2-(trifluoromethoxy)ethoxy]benzene
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1820736-26-2
:
Formula:C₉H₅F₆IO₂
MDL Number:MFCD28096622
:386.03 g/mol
Boiling Pt:53.4 °C (0.15 mm Hg)
:GHS07
Storage Temperature:Ambient
Frequently Bought Together