3-(4-Fluorophenyl)pentanedioic acid 95+%
Catalog # APOSPC48529-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:226.201 g/mol
EINECS:222-373-0
:GHS07
Storage Temperature:Refrigerator
CAS Number:3449-63-6
Formula:C₁₁H₁₁FO₄
MDL Number:MFCD19352347