4-Bromo-3,3,4,4-tetrafluorobutyl tert-butyl carbonate
Catalog # APOSPC450469-25G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:325.095 g/mol
Storage Temperature:Ambient
CAS Number:1980085-12-8
Formula:C₉H₁₃BrF₄O₃
MDL Number:MFCD28334474