4-Bromo-3,3,4,4-tetrafluorobutyl butyl carbonate
Catalog # APOSPC450456-25G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:325.095 g/mol
Storage Temperature:Ambient
CAS Number:1923065-51-3
Formula:C₉H₁₃BrF₄O₃
MDL Number:MFCD28334473