4-Bromo-3,3,4,4-tetrafluorobutyl isopropyl carbonate
Catalog # APOSPC450443-25G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:311.069 g/mol
Storage Temperature:Ambient
CAS Number:1980035-20-8
Formula:C₈H₁₁BrF₄O₃
MDL Number:MFCD28334472