4-Bromo-3,3,4,4-tetrafluorobutyl propyl carbonate
Catalog # APOSPC450430-25G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:311.069 g/mol
Storage Temperature:Ambient
CAS Number:1923267-13-3
Formula:C₈H₁₁BrF₄O₃
MDL Number:MFCD28334471