4-Bromo-3,3,4,4-tetrafluorobutyl methyl carbonate
Catalog # APOSPC450406-25G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:283.016 g/mol
Storage Temperature:Ambient
CAS Number:1923065-47-7
Formula:C₆H₇BrF₄O₃
MDL Number:MFCD28334469