Order Entry
United Kingdom
ContactUsLinkComponent
4-Bromo-3,3,4,4-tetrafluorobutyl 1H,1H,7H-perfluoroheptyl carbonate
4-Bromo-3,3,4,4-tetrafluorobutyl 1H,1H,7H-perfluoroheptyl carbonate
Catalog # APOSPC450395-25G
Supplier:  Apollo Scientific
CAS Number:  
4-Bromo-3,3,4,4-tetrafluorobutyl 1H,1H,7H-perfluoroheptyl carbonate
Catalog # APOSPC450395-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :583.061 g/mol
Storage Temperature:Ambient
CAS Number:1980035-16-2
Formula:C₁₂H₇BrF₁₆O₃
MDL Number:MFCD28334467