Order Entry
United Kingdom
ContactUsLinkComponent
4-Bromo-3,3,4,4-tetrafluorobutyl 1H,1H-perfluorohexyl carbonate
4-Bromo-3,3,4,4-tetrafluorobutyl 1H,1H-perfluorohexyl carbonate
Catalog # APOSPC450393-25G
Supplier:  Apollo Scientific
CAS Number:  
4-Bromo-3,3,4,4-tetrafluorobutyl 1H,1H-perfluorohexyl carbonate
Catalog # APOSPC450393-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :551.044 g/mol
Storage Temperature:Ambient
CAS Number:1980035-06-0
Formula:C₁₁H₆BrF₁₅O₃
MDL Number:MFCD28334465