Order Entry
United Kingdom
ContactUsLinkComponent
4-Bromo-3,3,4,4-tetrafluorobutyl 2,2,3,3,3-pentafluoropropyl carbonate
4-Bromo-3,3,4,4-tetrafluorobutyl 2,2,3,3,3-pentafluoropropyl carbonate
Catalog # APOSPC450386-25G
Supplier:  Apollo Scientific
CAS Number:  
4-Bromo-3,3,4,4-tetrafluorobutyl 2,2,3,3,3-pentafluoropropyl carbonate
Catalog # APOSPC450386-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :401.021 g/mol
Storage Temperature:Ambient
CAS Number:1923267-05-3
Formula:C₈H₆BrF₉O₃
MDL Number:MFCD28334458