4-Bromo-3,3,4,4-tetrafluorobutyl hexafluoroisopropyl carbonate
Catalog # APOSPC450385-25G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:419.012 g/mol
Storage Temperature:Ambient
CAS Number:1980085-10-6
Formula:C₈H₅BrF₁₀O₃
MDL Number:MFCD28334457