4-Bromo-3,3,4,4-tetrafluorobutyl 2,2-difluoroethyl carbonate
Catalog # APOSPC450384-25G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:333.023 g/mol
Storage Temperature:Ambient
CAS Number:1980034-94-3
Formula:C₇H₇BrF₆O₃
MDL Number:MFCD28334456