Order Entry
United Kingdom
ContactUsLinkComponent
4-Bromo-3,3,4,4-tetrafluorobutyl 2,2,2-trifluoroethyl carbonate
4-Bromo-3,3,4,4-tetrafluorobutyl 2,2,2-trifluoroethyl carbonate
Catalog # APOSPC450383-25G
Supplier:  Apollo Scientific
CAS Number:  
4-Bromo-3,3,4,4-tetrafluorobutyl 2,2,2-trifluoroethyl carbonate
Catalog # APOSPC450383-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :351.014 g/mol
Storage Temperature:Ambient
CAS Number:1980035-19-5
Formula:C₇H₆BrF₇O₃
MDL Number:MFCD28334455