[1H,1H-Perfluoro(2,5-dimethyl-3,6-dioxanonanoyl)] acrylate
Catalog # APOSPC450143-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:536.139 g/mol
Storage Temperature:Ambient
CAS Number:17559-01-2
Formula:C₁₂H₅F₁₇O₄
MDL Number:MFCD16621322