Order Entry
United Kingdom
ContactUsLinkComponent
(1R)-1-[3,5-Bis(trifluoromethyl)phenyl]-N-[(1R)-1-phenylethyl]ethanaminium 2-carboxybenzoate
(1R)-1-[3,5-Bis(trifluoromethyl)phenyl]-N-[(1R)-1-phenylethyl]ethanaminium 2-carboxybenzoate
Catalog # APOSPC450032-250MG
Supplier:  Apollo Scientific
CAS Number:  
(1R)-1-[3,5-Bis(trifluoromethyl)phenyl]-N-[(1R)-1-phenylethyl]ethanaminium 2-carboxybenzoate
Catalog # APOSPC450032-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :527.456 g/mol
Storage Temperature:Ambient
Formula:C₁₈H₁₈F₆N·C₈H₅O₄
MDL Number:MFCD28397827