Order Entry
United Kingdom
ContactUsLinkComponent
 
3-Fluoro-4-nitrophenylacetonitrile 95%
Catalog # APOSPC4405-250MG
Supplier:  Apollo Scientific
CAS Number:  
3-Fluoro-4-nitrophenylacetonitrile 95%
Catalog # APOSPC4405-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590
  • Environmentally Preferable:

Specifications

About this item

 :
 :180.136 g/mol
Storage Temperature:Ambient
CAS Number:503315-75-1
Formula:C₈H₅FN₂O₂
MDL Number:MFCD03094238