Order Entry
United Kingdom
ContactUsLinkComponent
2-(3-Fluorophenoxy)-6-chloropyrazine
2-(3-Fluorophenoxy)-6-chloropyrazine
Catalog # APOSPC430567-250MG
Supplier:  Apollo Scientific
CAS Number:  
2-(3-Fluorophenoxy)-6-chloropyrazine
Catalog # APOSPC430567-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :224.619 g/mol
Storage Temperature:Ambient
CAS Number:894416-95-6
Formula:C₁₀H₆ClFN₂O
MDL Number:MFCD09952090