6-Chloro-n-(4-fluorophenyl)pyridin-3-amine
Catalog # APOSPC430560-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:222.646 g/mol
Storage Temperature:Ambient
CAS Number:509953-60-0
Formula:C₁₁H₈ClFN₂
MDL Number:MFCD16987532