3-Fluoro-4-phenoxyaniline hydrochloride
Catalog # APOSPC410113-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:239.673 g/mol
Storage Temperature:Ambient
CAS Number:1431963-13-1
Formula:C₁₂H₁₀FNO·ClH
MDL Number:MFCD25371195