Order Entry
United Kingdom
ContactUsLinkComponent
3-Chloro-4-[3-(trifluoromethyl)-4,5,6,7-tetrahydro-1H-indazol-1-yl]aniline hydrochloride
3-Chloro-4-[3-(trifluoromethyl)-4,5,6,7-tetrahydro-1H-indazol-1-yl]aniline hydrochloride
Catalog # APOSPC410080-1G
Supplier:  Apollo Scientific
CAS Number:  
3-Chloro-4-[3-(trifluoromethyl)-4,5,6,7-tetrahydro-1H-indazol-1-yl]aniline hydrochloride
Catalog # APOSPC410080-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :352.182 g/mol
Storage Temperature:Ambient
Formula:C₁₄H₁₃ClF₃N₃·ClH
MDL Number:MFCD25371103