3-Fluoro-4-(1H-pyrazol-1-yl)aniline hydrochloride
Catalog # APOSPC410077-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:213.639 g/mol
Storage Temperature:Ambient
Formula:C₉H₈FN₃·ClH
MDL Number:MFCD12197237