Order Entry
United Kingdom
ContactUsLinkComponent
4-[5-Cyclopropyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]aniline hydrochloride
4-[5-Cyclopropyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]aniline hydrochloride
Catalog # APOSPC410039-1G
Supplier:  Apollo Scientific
CAS Number:  
4-[5-Cyclopropyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]aniline hydrochloride
Catalog # APOSPC410039-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :303.711 g/mol
Storage Temperature:Ambient
Formula:C₁₃H₁₂F₃N₃·ClH
MDL Number:MFCD25371072