Order Entry
United Kingdom
ContactUsLinkComponent
2-[(4-Fluoro-benzenesulphonylamino)-methyl]-piperidine-1-carboxylic acid tert-butyl ester
2-[(4-Fluoro-benzenesulphonylamino)-methyl]-piperidine-1-carboxylic acid tert-butyl ester
Catalog # APOSPC408377-500MG
Supplier:  Apollo Scientific
CAS Number:  
2-[(4-Fluoro-benzenesulphonylamino)-methyl]-piperidine-1-carboxylic acid tert-butyl ester
Catalog # APOSPC408377-500MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :372.455 g/mol
Storage Temperature:Ambient
CAS Number:1353980-93-4
Formula:C₁₇H₂₅FN₂O₄S
MDL Number:MFCD21098973