3-Fluoro-N-(S)-pyrrolidin-3-yl-benzenesulphonamide hydrochloride
Catalog # APOSPC408374-500MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:500 mg
- Seq:2725
Specifications
About this item
:
:280.747 g/mol
Storage Temperature:Ambient
CAS Number:1354018-70-4
Formula:C₁₀H₁₃FN₂O₂S·ClH
MDL Number:MFCD21098947