2-(4-Fluoro-phenylsulphanylmethyl)-pyrrolidine hydrochloride
Catalog # APOSPC408347-500MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:500 mg
- Seq:2725
Specifications
About this item
:
:247.76 g/mol
Storage Temperature:Ambient
CAS Number:1353989-69-1
Formula:C₁₁H₁₄FNS·ClH
MDL Number:MFCD21098493