(R)-3-(4-Fluoro-phenylsulphanyl)-pyrrolidine hydrochloride
Catalog # APOSPC408330-500MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:500 mg
- Seq:2725
Specifications
About this item
:
:233.733 g/mol
Storage Temperature:Ambient
CAS Number:1289584-92-4
Formula:C₁₀H₁₂FNS·ClH
MDL Number:MFCD18837555