4-(4-Fluoro-phenylsulphanylmethyl)-piperidine hydrochloride
Catalog # APOSPC408324-500MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:500 mg
- Seq:2725
Specifications
About this item
:
:261.786 g/mol
Storage Temperature:Ambient
CAS Number:1289384-72-0
Formula:C₁₂H₁₆FNS·ClH
MDL Number:MFCD18837521