5-Fluoro-2-(pyrrolidin-2-ylmethoxy)-pyrimidine hydrochloride
Catalog # APOSPC408315-500MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:500 mg
- Seq:2725
Specifications
About this item
:
:233.67 g/mol
Storage Temperature:Ambient
CAS Number:1261235-88-4
Formula:C₉H₁₂FN₃O·ClH
MDL Number:MFCD18380140