(S)-3-(4-Fluoro-benzenesulphinyl)-pyrrolidine hydrochloride
Catalog # APOSPC408298-500MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:500 mg
- Seq:2725
Specifications
About this item
:
:249.733 g/mol
Storage Temperature:Ambient
CAS Number:1354025-28-7
Formula:C₁₀H₁₂FNOS·ClH
MDL Number:MFCD21098442