5-Bromo-2-(4-methylpiperazin-1-yl)-3-(trifluoromethyl)pyridine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1208081-84-8
:
Formula:C₁₁H₁₃BrF₃N₃
MDL Number:MFCD15142779
:324.14 g/mol
Storage Temperature:Ambient
Frequently Bought Together