Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United Kingdom
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Nitro-1,1,1-trifluoropropan-2-ol
3-Nitro-1,1,1-trifluoropropan-2-ol 97%
3-Nitro-1,1,1-trifluoropropan-2-ol 97%
3-Nitro-1,1,1-trifluoropropan-2-ol 97%
APOSPC32425-25G
:
Apollo Scientific
:
3-Nitro-1,1,1-trifluoropropan-2-ol 97%
APOSPC32425-25G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
25 g
Seq:
3308
Environmentally Preferable:
:
2-Hydroxy-1-nitro-3,3,3-trifluoropropane
:
:
159.064 g/mol
:
Ambient
:
453-35-0
:
C₃H₄F₃NO₃
:
MFCD01328287