3-Chloro-2-fluoro-5-(trifluoromethoxy)benzoic acid 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:2385608-20-6
:
Formula:C₈H₃ClF₄O₃
MDL Number:MFCD32689357
:258.554 g/mol
Storage Temperature:Ambient
Frequently Bought Together