2-Bromo-5-fluoro-4-(trifluoromethyl)benzenesulphonyl chloride
Catalog # APOSPC303561-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:341.505 g/mol
Storage Temperature:Ambient
CAS Number:2383916-69-4
Formula:C₇H₂BrClF₄O₂S
MDL Number:MFCD32640139