Order Entry
United Kingdom
ContactUsLinkComponent
 
5-Bromo-2,3-difluorobenzenesulphonyl chloride
Catalog # APOSPC303431-1G
Supplier:  Apollo Scientific
CAS Number:  
5-Bromo-2,3-difluorobenzenesulphonyl chloride
Catalog # APOSPC303431-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :291.498 g/mol
Storage Temperature:Ambient
CAS Number:1805119-83-8
Formula:C₆H₂BrClF₂O₂S
MDL Number:MFCD28737881