4-Chloro-3-ethoxy-2-fluorobenzenesulphonyl chloride
Catalog # APOSPC303268-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:273.109 g/mol
Storage Temperature:Ambient
CAS Number:1706430-95-6
Formula:C₈H₇Cl₂FO₃S
MDL Number:MFCD28054241