3-Bromo-5-(trifluoromethoxy)benzenesulphonyl chloride
Catalog # APOSPC303248-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:339.514 g/mol
Storage Temperature:Ambient
CAS Number:1706446-78-7
Formula:C₇H₃BrClF₃O₃S
MDL Number:MFCD28054230