3-Chloro-4-ethoxy-5-fluorobenzoyl chloride
Catalog # APOSPC302714-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:237.055 g/mol
Storage Temperature:Ambient
CAS Number:1017778-76-5
Formula:C₉H₇Cl₂FO₂
MDL Number:MFCD09832337