4-Fluoro-3-(trifluoromethoxy)benzoyl chloride 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:886501-08-2
:
Formula:C₈H₃ClF₄O₂
MDL Number:MFCD06660252
:242.555 g/mol
Storage Temperature:Ambient
Frequently Bought Together