Order Entry
United Kingdom
ContactUsLinkComponent
4-Chloro-6-{4-[3-(trifluoromethyl)phenyl]piperazin-1-yl}pyrimidin-2-amine
4-Chloro-6-{4-[3-(trifluoromethyl)phenyl]piperazin-1-yl}pyrimidin-2-amine
Catalog # APOSPC300961-1G
Supplier:  Apollo Scientific
CAS Number:  
4-Chloro-6-{4-[3-(trifluoromethyl)phenyl]piperazin-1-yl}pyrimidin-2-amine
Catalog # APOSPC300961-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047
  • Environmentally Preferable:

Specifications

About this item

 :
 :357.761 g/mol
Storage Temperature:Ambient
CAS Number:2183593-91-9
Formula:C₁₅H₁₅ClF₃N₅
MDL Number:MFCD29060040